missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Lactulose Standard for IC™, 1,000 mg/L in Water, MilliporeSigma™ Supelco™

Catalog No. 111004353
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004353 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004353 Supplier Merck Emd Millipore Supplier No. 0144850ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.

Specifications

Product Line TraceCERT
Purity Grade Notes 1000 mg/L lactulose aq. soln., prepared with lactulose and water; filtered through a 0.2 mmu membrane filter
Concentration 1000 mg/L aq. soln.
Type Certified Reference Material
Quantity 50 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC