missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ L-Methionine sulfoximine, MilliporeSigma™ Supelco™

Catalog No. 111013274
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013274 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013274 Supplier Merck Emd Millipore Supplier No. 91016100MG
Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 15985-39-4
Physical Form Neat
Linear Formula CH3 S(O)(=NH)CH2 CH2 CH(NH2)CO2 H
MDL Number MFCD00002621
Melting Point >210°C
Synonym L-S-(3-Amino-3-carboxypropyl)-S-methylsulfoximine
Type Certified Reference Material
Quantity 100 mg
Formula Weight 180.23
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC