missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Imidacloprid, MilliporeSigma™ Supelco™

Catalog No. 111018543
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018543 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018543 Supplier Merck Emd Millipore Supplier No. 6869450MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 138261-41-3
Product Line TraceCERT
Linear Formula C9H10 ClN5O2
MDL Number MFCD00468059
Type Certified Reference Material
Quantity 50 mg
Formula Weight 255.66
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC