missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ HPLC Isocratic Systems Diagnostics Mix in Methanol:Water (3:2), MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 67-56-1 |
| Flash Point | 9°C |
| Type | Certified Reference Material |
| Quantity | 6 x 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC