missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ (-)-Guaiol, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Product Line | TraceCERT |
| Flash Point | 9.7°C - closed cup |
| Concentration | 2,000 μg/mL in methanol |
| Synonym | Cannabis terpenes; Terpenes |
| Type | Pharmaceuticals, Illicit Drugs and Alcohol Standards |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC