missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Glucose Standard for IC, 1,000 mg/L in Water, MilliporeSigma™ Supelco™
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.
Specifications
Specifications
| Product Line | TraceCERT |
| Purity Grade Notes | 1000 mg/L glucose aq. soln., prepared with glucose and water; filtered through a 0.2 mum membrane filter |
| Concentration | 1000 mg/L aq. soln. |
| Type | Certified Reference Material |
| Quantity | 50 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC