missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ (±)-Geosmin, 100 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017883
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017883 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017883 Supplier Merck Emd Millipore Supplier No. CRM47522
Only null left

Specifications

Packaging Ampule
CAS 16423-19-1
Product Line TraceCERT
Flash Point 9.7°C - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C12H22 O
Synonym Drinking Water Odor Standards; Odor Standards
Type Certified Reference Material
Quantity 1 mL
Formula Weight 182.30
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC