missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ F.A.M.E. Mix RM-1, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111020919
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020919 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020919 Supplier Merck Emd Millipore Supplier No. O70061AMP
Only null left

Our quantitative animal and vegetable reference mixes conform to the requirements of the American Oil Chemists' Society (AOCS) method Ce 1-62. The composition of each mix is similar to the fatty acid distribution of certain oils listed under each product. Please note that the AOCS mixtures sold under the Supelco brand are named RM-1, RM-2, etc.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Physical Form Neat
Synonym AOCS Reference Mixtures; AOCS No. 1 (RM-1); Oil Reference Standard, AOCS No. 1; Oil Reference Standard, AOCS
Quantity 100 mg
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Methyl arachidate 3 wt. %; Methyl linoleate 50 wt. %; Methyl linolenate 3 wt. %; Methyl oleate 35 wt. %; Methyl palmitate 6 wt. %; Methyl stearate 3 wt. %

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC