missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ F.A.M.E. Mix C8 - C22 (Neat), MilliporeSigma™ Supelco™

Catalog No. 111002574
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002574 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002574 Supplier Merck Emd Millipore Supplier No. CRM18920

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 111-82-0
Product Line TraceCERT
Shipping Condition Dry Ice
Synonym Fatty Acid Methyl Ester Mix
Type Certified Reference Material
Quantity 100 mg
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC