missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ F.A.M.E. Column Evaluation Mix, 1,000 μg/mL each component in dichloromethane, MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 75-09-2 |
| Physical Form | Liquid |
| Type | Certified Reference Material |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 μg/mL each component in dichloromethane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC