missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Etofenprox certified reference material, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Product Line | TraceCERT |
| CAS | 80844-07-1 |
| Linear Formula | C25H28O3 |
| MDL Number | MFCD00210287 |
| Synonym | 2-(4-Ethoxyphenyl)-2-methylpropyl 3-phenoxybenzyl ether |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 376.49 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC