missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Ethylene oxide Solution, 50 mg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020050
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020050 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020050 Supplier Merck Emd Millipore Supplier No. 48838

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 75-21-8
Flash Point 10°C
Linear Formula C2H4 O
MDL Number MFCD00014482
Type Food and Cosmetic Component Standard
Quantity 1 mL
Formula Weight 44.05
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 50 mg/mL in methanol
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC