missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Epicatechin Gallate Solution, Certified Reference Material, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL in methanol |
| Quantity | 0.5 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC