missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ EPA Phthalate Esters Mix, 2,000 μg/mL each component in Methanol, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 67-56-1 |
| Synonym | EPA Phthalate Esters Mix |
| Type | Certified Reference Material |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL each component in methanol |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC