missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ EPA CLP Organochlorine Pesticide MIx, 2,000 μg/mL Each Component in Hexane: Toluene (1:1), MilliporeSigma™ Supelco™

Catalog No. 111017369
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017369 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017369 Supplier Merck Emd Millipore Supplier No. CRM47426
Only null left

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 2,000 μg/mL each component in hexane: toluene (1:1)
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) alpha-BHC; beta-BHC; Lindane; delta-BHC; Heptachlor; Aldrin; Heptachlorepoxide Isomer B; gamma-Chlordane; alpha-Chlordane; Endosulfan I (alpha); 4,4 '-DDE; Dieldrin; Endrin; Endosulfan II (Beta Isomer); 4,4 '-DDD; Endrin aldehyde; Endosulfan sulfate; 4,4 '-DDT; Endrin Ketone; Methoxychlor

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC