missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ EPA 8270 Semivolatile Internal Standard Mix, 2,000 μg/mL, MilliporeSigma™ Supelco™
2,000 μg/mL Each Component in Methylene Chloride
$101.56 - $101.56
Specifications
| Packaging | Ampule |
|---|---|
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL each component in methylene chloride |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Acenaphthene-d10; Chrysene-d12; 1,4-Dichlorobenzene-d4; Naphthalene-d8; Perylene-d12; Phenanthrene-d10 |
Description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.Specifications
| Ampule | |
| 2,000 μg/mL each component in methylene chloride | |
| Acenaphthene-d10; Chrysene-d12; 1,4-Dichlorobenzene-d4; Naphthalene-d8; Perylene-d12; Phenanthrene-d10 |
| TraceCERT | |
| TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC