missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ EPA 8270 Organophosphorus Pesticide Mix 2, 2,000 μg/mL Each Component in Dichloromethane, MilliporeSigma™ Supelco™

Catalog No. 111017253
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017253 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017253 Supplier Merck Emd Millipore Supplier No. CRM47908

Includes

Ampule of 1 mL

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Description Quantity: 1 mL
Packaging Ampule of 1 mL
Product Line TraceCERT
Concentration 2,000 μg/mL each component in dichloromethane
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Dimethoate; Disulfoton; Famphur; Parathion; Parathion-methyl; Phorate; Sulfotep; Thionazin; Triethyl thiophosphate

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC