missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ EPA 8260 Internal Standards Mix HC, 10,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017126
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017126 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017126 Supplier Merck Emd Millipore Supplier No. 47082U
Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 67-56-1
Flash Point 50°C
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 10,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC