missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ EPA 606-M Phthalate Esters Mix, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022580
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022580 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022580 Supplier Merck Emd Millipore Supplier No. 48741

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

Chemical Name or Material Organic Pollutant Standards
Flash Point 9.7°C (Closed Cup)
Packaging Ampule
Quantity 1 mL
UN Number UN1230 - Class 3 - PG 2 - Methanol, solution
Grade Trace CERT

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC