missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ EPA 501/601 Trihalomethanes Calibration Mix, 200 μg/mL Each Component in Methanol, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 200 μg/mL each component in methanol |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Bromodichloromethane; Bromoform; Chloroform; Dibromochloromethane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC