missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Elemental Impurities according to ICH Q3D Inhalation, Standard 2, MilliporeSigma™ Supelco™

Catalog No. 111016911
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016911 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016911 Supplier Merck Emd Millipore Supplier No. 93696100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.Certified content incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 7647-01-0
Product Line TraceCERT
Purity Grade Notes The concentrations are 10 fold according to ICH Q3 D guideline.
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Pd, 10 mg/L

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC