missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Dicofol certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018447
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018447 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018447 Supplier Merck Emd Millipore Supplier No. 7404650MG
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

Product Line TraceCERT
CAS 115-32-2
Linear Formula C14H9 Cl5 O
MDL Number MFCD00055271
Synonym 2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol
Type Certified Reference Material
Quantity 50 mg
Formula Weight 370.49
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC