missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Dichloromethane Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020308
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020308 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020308 Supplier Merck Emd Millipore Supplier No. 40042

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture and Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 75-09-2
Density 0.791 g/cm3 (at 20°C)
Linear Formula CH2 Cl2
MDL Number MFCD00000881
Synonym Methylene chloride solution
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 84.93
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 5,000 μg/mL in methanol
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC