missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ DFTPP Solution, 1,000 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111020433
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020433 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020433 Supplier Merck Emd Millipore Supplier No. 47941
Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 5074-71-5
Synonym Decafluorotriphenylphosphine solution; 2,3,4,5,6,2 ′,3 ′,4 ′,5 ′,6 ′-Decafluorotriphenylphosphine; Bis(pentafluorophenyl)phenylphosphine
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 μg/mL in acetone

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC