missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ λ-Cyhalothrin Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019172
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019172 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019172 Supplier Merck Emd Millipore Supplier No. 7276550MG
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

Product Line TraceCERT
CAS 91465-08-6
Physical Form Neat
Linear Formula C23H19 ClF3 NO3
MDL Number MFCD07370148
Type Certified Reference Material
Quantity 50 mg
Formula Weight 449.85
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC