missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Cyanide Standard for IC, 1,000 mg/L Cyanide in Water, MilliporeSigma™ Supelco™

Catalog No. 111020021
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020021 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020021 Supplier Merck Emd Millipore Supplier No. 90157100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.

Specifications

CAS 14244-62-3
Product Line TraceCERT
Purity Grade Notes 1000 mg/L cyanide aq. soln., prepared with high purity K2 [Zn(CN)4 ] and water. “1000 mg/L CN- aq. soln.” refers to the nominal cyanide-concentration and not to the form of the analyte in the solution. Cyanide exists mainly in the complex form ([Zn(CN)4]2-) in the water (basic) solution.
Concentration 1000 mg/L cyanide aq. soln.
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC