missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Citronellol Solution, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017875
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017875 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017875 Supplier Merck Emd Millipore Supplier No. CRM40469

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Food and Beverages and Pharmaceutical (small molecule)

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 106-22-9
Flash Point 9.7°C - Closed Cup
Type Pharmaceuticals, Illicit Drugs and Alcohol Standards
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC