missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Citric acid, MilliporeSigma™ Supelco™

Catalog No. 111015530
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015530 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015530 Supplier Merck Emd Millipore Supplier No. 94676100MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 77-92-9
Physical Form Neat
Linear Formula HOC(COOH)(CH2 COOH)2
MDL Number MFCD00011669
Melting Point 153°C to 159°C
Type Certified Reference Material
Quantity 100 mg
Formula Weight 192.12
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC