missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Calcium Formate Standard, MilliporeSigma™ Supelco™

Catalog No. 111014555
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014555 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014555 Supplier Merck Emd Millipore Supplier No. 038261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium formate is a water soluble salt, which can be obtained as a by-product in the manufacture of polyhydric alcohol. It can serve as an efficient source of calcium in dietary supplements.

Specifications

Description qNMR Standard for D2O (7.5 - 8.5 ppm)
CAS 544-17-2
Product Line TraceCERT
Linear Formula (HCOO)2 Ca
MDL Number MFCD00036108
Synonym Formic acid calcium salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 130.11
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC