missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Butylated hydroxyanisole, MilliporeSigma™ Supelco™

Catalog No. 111013697
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013697 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013697 Supplier Merck Emd Millipore Supplier No. 4988650MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line TraceCERT
Ignition Point 599°F
CAS 25013-16-5
Vapor Density 6.2 (vs. air)
Flash Point 113°C - Closed Cup
Linear Formula (CH3)3 CC6H3(OCH3)OH
MDL Number MFCD01779059
Melting Point 58°C to 60°C
Synonym 2(3)-t-Butyl-4-hydroxyanisole; 2(3)-t-Butylhydroquinone monomethyl ether; BHA
Type Certified Reference Material
Quantity 50 mg
Formula Weight 180.24
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC