missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Bromomethane Solution, 200 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020570
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020570 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020570 Supplier Merck Emd Millipore Supplier No. 48624

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bromomethane is frequently used as fumigant. It is a liquid at room temperature which readily vaporizes into a colourless gas. It is slightly soluble in water and is freely soluble in organic solvents.

Agriculture and Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 74-83-9
Flash Point 11°C
Linear Formula CH3 Br
MDL Number MFCD00000166
Synonym Methyl bromide
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 94.94
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 200 μg/mL in methanol
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC