missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Bromodichloromethane Solution, 200 μg/mL in Methanol, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 75-27-4 |
| Flash Point | 11°C |
| Linear Formula | CHBrCl2 |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 163.83 |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 200 μg/mL in methanol |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC