missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Bromate Standard for IC, 1,000 mg/L Bromate in Water, MilliporeSigma™ Supelco™

Catalog No. 111002658
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002658 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002658 Supplier Merck Emd Millipore Supplier No. 78476100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1-2-7758
Product Line TraceCERT
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 mg/L bromate aq. soln.

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC