missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Boscalid, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022770
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022770 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022770 Supplier Merck Emd Millipore Supplier No. 7414450MG

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 188425-85-6
Molecular Formula C18H12Cl2N2O
Linear Formula C18H12Cl2N2O
MDL Number MFCD06795150
Synonym 2-Chloro-N-(4'-chlorobiphenyl-2- yl)-nicotinamide
Quantity 50 mg
Formula Weight 343.21
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC