missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Boron Standard for AAS, 1,000 mg/L B in Water, MilliporeSigma™ Supelco™

Catalog No. 111002656
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002656 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002656 Supplier Merck Emd Millipore Supplier No. 40591250ML

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.

Specifications

Product Line TraceCERT
CAS 10043-35-3
Type Certified Reference Material
Quantity 250 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 mg/L B aq. soln.

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC