missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Bis(2-ethylhexyl) phthalate Solution, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 117-81-7 |
| Flash Point | 11°C |
| Linear Formula | C24H38O4 |
| Type | Food and Cosmetic Component Standard |
| Quantity | 1 mL |
| Formula Weight | 390.56 |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL in methanol |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC