missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ δ-BHC, 1,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017899
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017899 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017899 Supplier Merck Emd Millipore Supplier No. 40103U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 319-86-8
Flash Point 11°C
Linear Formula C6H6 Cl6
Synonym delta-HCH solution; delta-1,2,3,4,5,6-Hexachlorocyclohexane solution
Type Certified Reference Material
Quantity 1 mL
Formula Weight 290.83
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 μg/mL in methanol
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC