missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Aroclor 1254 Solution, 50 mg/kg in transformer oil, MilliporeSigma™ Supelco™

Catalog No. 111019108
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019108 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019108 Supplier Merck Emd Millipore Supplier No. 48734
Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 11097-69-1
Type Organic Pollutant Standard
Quantity 5 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 50 mg/kg in transformer oil

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC