missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Anthracened10 Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™

Catalog No. 111020358
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020358 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020358 Supplier Merck Emd Millipore Supplier No. SGMA48863
Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 1719-06-8
Linear Formula C14D10
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 188.29
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in dichloromethane

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC