missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Anhydrotetracycline hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013022
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013022 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013022 Supplier Merck Emd Millipore Supplier No. 9466410MG
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Pharmaceutical (small molecule)

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 13803-65-1
Shipping Condition Wet ice
Physical Form Neat
Linear Formula C22H22N2O7 · HCl
MDL Number MFCD00151453
Synonym (4 S,4 AS,12 AS)-4-(Dimethylamino)-1,4,4 A,5,12,12 A-hexahydro-3,10,11,12 A-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride
Type Certified Reference Material
Quantity 10 mg
Formula Weight 462.88
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC