missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ γ-Aminobutyric acid, MilliporeSigma™ Supelco™

Catalog No. 111014622
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014622 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014622 Supplier Merck Emd Millipore Supplier No. 4381150MG

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Food and Beverages

Specifications

Product Line TraceCERT
CAS 56-12-2
Linear Formula NH2(CH2)3 COOH
MDL Number MFCD00008226
Melting Point 195°C
Synonym 3-Carboxypropylamine; 4-Aminobutanoic acid; GABA; Piperidic acid; Piperidinic acid
Type Certified Reference Material
Quantity 50 mg
Formula Weight 103.12
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC