missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Alpha-Amylcinnamyl Alcohol, 2000 μg/mL, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 101-85-9 |
| Flash Point | 9.7°C (Closed Cup) |
| Concentration | 2000 μg/mL in methanol |
| UN Number | UN1230 - Class 3 - PG 2 - Methanol, solution |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC