missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ all-cis-4, 7, 10, 13, 16, 19-Docosahexaenoic Acid methyl ester, 10 mg/mL in Heptane, MilliporeSigma™ Supelco™

Catalog No. 111016609
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016609 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016609 Supplier Merck Emd Millipore Supplier No. CRM47570

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 142-82-5
Product Line TraceCERT
Flash Point -3.99°C - closed cup
Physical Form Liquid
Concentration 10 mg/mL in heptane
Type Certified Reference Material
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC