missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Acetone-2,4-DNPH, MilliporeSigma™ Supelco™

Catalog No. 111013253
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013253 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013253 Supplier Merck Emd Millipore Supplier No. 0137850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material TraceCERT™ suitable for quantitative analysis.

Specifications

Chemical Name or Material Acetone-2,4-dinitrophenylhydrazone
CAS 1567-89-1
Product Line TraceCERT
Linear Formula (CH3)2 C=NNHC6H3(NO2)2
Technique Gas Chromatography
MDL Number MFCD00137172
Certifications/Compliance ISO/IEC 17025
Melting Point 126°C to 128°C
Synonym Acetone-2,4-dinitrophenylhydrazone
Type Certified Reference Material
Solvent or Matrix Neat
Molecular Weight (g/mol) 238.20
Industry Type Biotech
Quantity 50 mg
Formula Weight 238.20
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC