missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Acenaphthene-d10 Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 15067-26-2 |
| Linear Formula | C12D10 |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 164.27 |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL in dichloromethane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC