missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ 4,4'-Difluorobenzophenone, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022417
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022417 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022417 Supplier Merck Emd Millipore Supplier No. 07563500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 345-92-6
Melting Point 102°C to 105°C (literature)
Flash Point 152°C (Pensky-Martens Closed Cup)
Molecular Formula C13H8F2O
Linear Formula (FC6H4)2 CO
Quantity 500 mg
MDL Number MFCD00000353
UN Number UN 3077 9 / PGIII
Formula Weight 218.20
Grade Trace CERT

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC