missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ 2-Bromo-1-chloropropane Solution, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020280
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020280 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020280 Supplier Merck Emd Millipore Supplier No. 48088
Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 3017-95-6
Linear Formula C3H6 BrCl
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 157.44
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC