missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ 2,6-Dichlorophenol Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020109
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020109 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020109 Supplier Merck Emd Millipore Supplier No. 40302

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture and Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 87-65-0
Flash Point 11°C
Linear Formula C6H4 Cl2 O
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 163.00
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 5,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC