missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ 1,1,2-Trichloro-1,2,2-trifluoroethane, 1,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 76-13-1 |
| Synonym | 1,1,2-Trichloro-1,2,2-trifluoroethane solution |
| Type | Food and Cosmetic Component Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 μg/mL in methanol |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC