missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™
Description
Ideal for lab or field applications
Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.
Calibration within container — zero contamination
- Traceable to NIST (National Institute of Standards and Technology)
- Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
- 100% compatible with all conductivity meters
- A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
- Includes an individual temperature chart and step-by-step calibration instructions
- Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
- Single use Eliminates external contamination concerns Each standard contains 100mL
Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12
Order Info
Each bottle contains 100mL
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | 6/Pk. |
| Conductivity | 10 μ℧ |
| Technique | Calibration of Conductivity Instrument |
| Color | Clear |
| pH | 7 to 9 |
| For Use With (Equipment) | Conductivity Meter |
| Solubility | Soluble in water |
| Solution Type | Calibration Solution |
| Type | Conductivity Standard |
| Show More |